missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Loteprednol etabonate

Catalog No. AC467700050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC467700050 5 g
AC467700010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC467700050 Supplier Thermo Scientific Chemicals Supplier No. 467700050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 82034-46-6
Molecular Formula C24H31ClO7

Specifications

Quantity 5 g
Solubility Solubility aq. soln.: insoluble.
Melting Point 226.0°C to 233.0°C
Formula Weight 466.96
Chemical Name or Material Loteprednol etabonate

RUO – Research Use Only