accessibility menu, dialog, popup

UserName

CH3NNOH

CAS RN 81998-04-1

IUPAC Name: {4'-methyl-[2,2'-bipyridin]-4-yl}methanol
Synonyms: {4'-methyl-[2,2'-bipyridin]-4-yl}methanol
Molecular Weight (g/mol): 200.24
Molecular Formula: C12H12N2O
InChi Key: GJCOKGHYIMLMPB-UHFFFAOYSA-N
SMILES: CC1=CC(=NC=C1)C1=NC=CC(CO)=C1
Grade 
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results