missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxymethyl-4'-methyl-2,2'-bipyridyl 97.0+%, TCI America™

Catalog No. H1347200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H1347200MG 200 mg
H13471G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H1347200MG Supplier TCI America Supplier No. H1347200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 81998-04-1
Molecular Formula C12H12N2O
Molecular Weight (g/mol) 200.24
MDL Number MFCD02916382
InChI Key GJCOKGHYIMLMPB-UHFFFAOYSA-N
Synonym 4-Hydroxymethyl-4′C-methyl-2,2′C-bipyridine, 4′C-Methyl-2,2′C-bipyridine-4-methanol
PubChem CID 2754750
IUPAC Name {4'-methyl-[2,2'-bipyridin]-4-yl}methanol
SMILES CC1=CC(=NC=C1)C1=NC=CC(CO)=C1

Specifications

Melting Point 121°C
Color White-Yellow
Quantity 200 mg
Formula Weight 200.24
Percent Purity ≥97.0% (T)
Grade Reagent
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxymethyl-4′-methyl-2,2′-bipyridyl