accessibility menu, dialog, popup

UserName

CH3OH

CAS RN 81936-33-6

IUPAC Name: 4-(4-propylcyclohexyl)phenol
Synonyms: 4-(4-propylcyclohexyl)phenol
Molecular Weight (g/mol): 218.34
Molecular Formula: C15H22O
InChi Key: AHAZEMSUUYFDMM-UHFFFAOYSA-N
SMILES: CCCC1CCC(CC1)C1=CC=C(O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results