missing translation for 'onlineSavingsMsg'
Learn More

4-(trans-4-Propylcyclohexyl)phenol 98.0+%, TCI America™

Catalog No. P161625G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P161625G 25 g
P1616100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P161625G Supplier TCI America Supplier No. P161625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 81936-33-6
Molecular Formula C15H22O
Molecular Weight (g/mol) 218.34
MDL Number MFCD00673751
InChI Key AHAZEMSUUYFDMM-UHFFFAOYSA-N
Synonym 1-Hydroxy-4-(trans-4-propylcyclohexyl)benzene
PubChem CID 3126231
IUPAC Name 4-(4-propylcyclohexyl)phenol
SMILES CCCC1CCC(CC1)C2=CC=C(C=C2)O

Specifications

Melting Point 138°C
Color White-Yellow
Quantity 25 g
Formula Weight 218.34
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(trans-4-Propylcyclohexyl)phenol