accessibility menu, dialog, popup

UserName

ClClClNHO

CAS RN 81927-55-1

IUPAC Name: benzyl 2,2,2-trichloroethanimidate
Synonyms: benzyl 2,2,2-trichloroethanimidate
Molecular Weight (g/mol): 252.52
Molecular Formula: C9H8Cl3NO
InChi Key: HUZCTWYDQIQZPM-UHFFFAOYSA-N
SMILES: ClC(Cl)(Cl)C(=N)OCC1=CC=CC=C1
Boiling Point 
  • (2)
  • (4)
  • (1)
Percent Purity 
  • (1)
  • (6)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (6)
  • (2)
  • (4)
  • (1)
  • (1)
  • (6)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)