missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 2,2,2-Trichloroacetimidate 97.0+%, TCI America™

Catalog No. B148325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B148325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B148325G Supplier TCI America Supplier No. B148325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 81927-55-1
Molecular Formula C9H8Cl3NO
Molecular Weight (g/mol) 252.52
MDL Number MFCD00000805
InChI Key HUZCTWYDQIQZPM-UHFFFAOYSA-N
Synonym benzyl 2,2,2-trichloroacetimidate, benzyl trichloroacetimidate, benzyltrichloroacetimidate, trichloroacetimidic acid benzyl ester, 2,2,2-trichloro-1-phenylmethoxy ethanimine, ethanimidic acid, 2,2,2-trichloro-, phenylmethyl ester, benzyl 2,2,2-trichloroethanecarboximidate, benzyl2,2,2-trichloroacetimidate, acmc-209pmn, benzyl trichloracetimidate
PubChem CID 144968
IUPAC Name benzyl 2,2,2-trichloroethanimidate
SMILES ClC(Cl)(Cl)C(=N)OCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 150°C
Quantity 25 g
Formula Weight 252.52
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzyl 2,2,2-Trichloroacetimidate
TSCA No
Recommended Storage Refrigerator