accessibility menu, dialog, popup

UserName

CAS RN 819058-34-9

IUPAC Name: 2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Synonyms: 2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Molecular Weight (g/mol): 237.08
Molecular Formula: C12H17BFNO2
InChi Key: AIXGNRNTXUKZLC-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(F)=C(N)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results