missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-fluorobenzeneboronic acid pinacol ester, 96%

Catalog No. AAH52349MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52349MD 250 mg
AAH5234903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52349MD Supplier Thermo Scientific Chemicals Supplier No. H52349MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 819058-34-9 | C12H17BFNO2 | 237.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 819058-34-9
Molecular Formula C12H17BFNO2
Molecular Weight (g/mol) 237.08
MDL Number MFCD09033884
InChI Key AIXGNRNTXUKZLC-UHFFFAOYSA-N
Synonym 2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-amino-3-fluorophenylboronic acid pinacol ester, 4-amino-3-fluorophenylboronic acid, pinacol ester, benzenamine, 2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-amino-3-fluorophenylboronicacidpinacolester, 4-amino-3-fluorophenylboronic acid,pinacol ester, 4-amino-3-fluorobenzeneboronic acid pinacol ester, 4-amino-3-fluorophenyl boronic acid pinacol ester, 2-3-fluoro-4-aminophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 17906172
IUPAC Name 2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
SMILES CC1(C)OB(OC1(C)C)C1=CC(F)=C(N)C=C1

Specifications

Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 237.08
Percent Purity 96%
Chemical Name or Material 4-Amino-3-fluorobenzeneboronic acid pinacol ester
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only