accessibility menu, dialog, popup

UserName

ClCH3H3CNH2CH3H3C

CAS RN 819-79-4

IUPAC Name: bis(propan-2-yl)azanium chloride
Synonyms: diisopropylazanium chloride
Molecular Weight (g/mol): 137.65
Molecular Formula: C6H16ClN
InChi Key: URAZVWXGWMBUGJ-UHFFFAOYSA-N
SMILES: [Cl-].CC(C)[NH2+]C(C)C
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)
pH 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results