missing translation for 'onlineSavingsMsg'
Learn More

Ionic Strength Adjustment Solution (1M), METTLER TOLEDO™

Catalog No. 01912569
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01912569 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01912569 Supplier METTLER TOLEDO Supplier No. 30232624

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For potentiometric measurements, including ion selective electrodes

  • Ionic Strength Adjustment Solution
  • For potentiometric measurements, including ion selective electrodes

Specifications

Chemical Name or Material DIPA-HCL-ISA 1 M
Color Colorless
CAS 819-79-4
Boiling Point 100°C
Molecular Formula C6H16ClN
Concentration 1 M
Molecular Weight (g/mol) 137.651
Packaging Glass Bottle
pH 11.3
Quantity 1 L