accessibility menu, dialog, popup

UserName

BNH3CH3CH3CCH3HOOH

CAS RN 815581-79-4

IUPAC Name: {2-[(2,2,6,6-tetramethylpiperidin-1-yl)methyl]phenyl}boronic acid
Synonyms: 2-[(2,2,6,6-tetramethylpiperidin-1-yl)methyl]phenylboronic acid
Molecular Weight (g/mol): 275.2
Molecular Formula: C16H26BNO2
InChi Key: DOZVPMCVRDTTJK-UHFFFAOYSA-N
SMILES: CC1(C)CCCC(C)(C)N1CC1=CC=CC=C1B(O)O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results