missing translation for 'onlineSavingsMsg'
Learn More

2-[(2,2,6,6-Tetramethyl-1-piperidyl)methyl]phenylboronic Acid (contains varying amounts of Anhydride) 98.0+%, TCI America™

Catalog No. T29081G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T29081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T29081G Supplier TCI America Supplier No. T29081G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 815581-79-4
Molecular Formula C16H26BNO2
Molecular Weight (g/mol) 275.199
InChI Key DOZVPMCVRDTTJK-UHFFFAOYSA-N
Synonym 2-[(2,2,6,6-Tetramethyl-1-piperidyl)methyl]benzeneboronic Acid
PubChem CID 11231172
IUPAC Name [2-[(2,2,6,6-tetramethylpiperidin-1-yl)methyl]phenyl]boronic acid
SMILES B(C1=CC=CC=C1CN2C(CCCC2(C)C)(C)C)(O)O

Specifications

Color White
Quantity 1 g
Formula Weight 275.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-[(2,2,6,6-Tetramethyl-1-piperidyl)methyl]phenylboronic Acid (contains varying amounts of Anhydride