accessibility menu, dialog, popup

UserName

CH3HNOFFF

CAS RN 815-06-5

IUPAC Name: 2,2,2-trifluoro-N-methylacetamide
Synonyms: 2,2,2-trifluoro-N-methylacetamide
Molecular Weight (g/mol): 127.07
Molecular Formula: C3H4F3NO
InChi Key: IQNHBUQSOSYAJU-UHFFFAOYSA-N
SMILES: CNC(=O)C(F)(F)F
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)