missing translation for 'onlineSavingsMsg'
Learn More

N-Methyltrifluoroacetamide 98.0+%, TCI America™

Catalog No. M081725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M081725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M081725G Supplier TCI America Supplier No. M081725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 815-06-5
Molecular Formula C3H4F3NO
Molecular Weight (g/mol) 127.066
MDL Number MFCD00009670
InChI Key IQNHBUQSOSYAJU-UHFFFAOYSA-N
Synonym n-methyltrifluoroacetamide, n-methyl-2,2,2-trifluoroacetamide, acetamide, 2,2,2-trifluoro-n-methyl, 2,2,2-trifluoro-n-methyl-acetamide, n-metnyl-2,2,2-trifluoroacetamide, methyltrifluoroacetamide, acmc-209plm, n-methyl-trifluoroacetamide, n-methyltrifluoro-acetamide
PubChem CID 69948
IUPAC Name 2,2,2-trifluoro-N-methylacetamide
SMILES CNC(=O)C(F)(F)F

Specifications

Melting Point 52°C
Color White
Boiling Point 160°C
Quantity 25 g
Formula Weight 127.07
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Methyltrifluoroacetamide