accessibility menu, dialog, popup

UserName

CAS RN 81270-37-3

H3COONHHCl

CAS RN 81270-37-3

IUPAC Name: methyl 2-(piperidin-4-yl)acetate hydrochloride
Synonyms: methyl 2-(piperidin-4-yl)acetate hydrochloride
Molecular Weight (g/mol): 193.67
Molecular Formula: C8H16ClNO2
InChi Key: ADBDFGZYGJGDNJ-UHFFFAOYSA-N
SMILES: Cl.COC(=O)CC1CCNCC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results