accessibility menu, dialog, popup

UserName

CAS RN 81172-89-6

IUPAC Name: 4-(diethoxymethyl)benzaldehyde
Synonyms: 4-(diethoxymethyl)benzaldehyde
Molecular Weight (g/mol): 208.26
Molecular Formula: C12H16O3
InChi Key: HTMXMFARWHNJDW-UHFFFAOYSA-N
SMILES: CCOC(OCC)C1=CC=C(C=O)C=C1
Percent Purity 
  • (1)
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)