missing translation for 'onlineSavingsMsg'
Learn More

Terephthalaldehyde Mono(diethyl Acetal) 97.0+%, TCI America™

Catalog No. T140125ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T140125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T140125ML Supplier TCI America Supplier No. T140125ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 81172-89-6
Molecular Formula C12H16O3
Molecular Weight (g/mol) 208.26
MDL Number MFCD00010217
InChI Key HTMXMFARWHNJDW-UHFFFAOYSA-N
Synonym 4-diethoxymethyl benzaldehyde, terephthalaldehyde mono diethyl acetal, benzaldehyde, 4-diethoxymethyl, terephthaldehyde mono diethyl acetal, terephthalaldehyde mono-diethyl acetal, pubchem22621, acmc-209tbs, 4-diethoxymethyl-benzaldehyde, 4-diethoxymethy benzaldehyde, 4-diethyoxymethyl benzaldehyde
PubChem CID 595993
IUPAC Name 4-(diethoxymethyl)benzaldehyde
SMILES CCOC(OCC)C1=CC=C(C=O)C=C1

Specifications

Color Yellow
Quantity 25 mL
Formula Weight 208.26
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Terephthalaldehyde Mono(diethyl Acetal)