accessibility menu, dialog, popup

UserName

CAS RN 81090-53-1

IUPAC Name: 4-bromo-N-(4-bromophenyl)-N-phenylaniline
Synonyms: 4-bromo-N-(4-bromophenyl)-N-phenylaniline
Molecular Weight (g/mol): 403.12
Molecular Formula: C18H13Br2N
InChi Key: KIGVOJUDEQXKII-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=C(Br)C=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)