missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dibromotriphenylamine 98.0+%, TCI America™

Catalog No. D19105G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D19105G 5 g
D19101G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D19105G Supplier TCI America Supplier No. D19105G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 81090-53-1
Molecular Formula C18H13Br2N
Molecular Weight (g/mol) 403.117
MDL Number MFCD00060104
InChI Key KIGVOJUDEQXKII-UHFFFAOYSA-N
Synonym 4,4'-dibromotriphenylamine, 4-bromo-n-4-bromophenyl-n-phenylaniline, n,n-bis 4-bromophenyl aniline, 4,4-dibromotriphenylamine, 4-bromo-n-4-bromophenyl-n-phenylbenzenamine, dibromotriphenylamine, 4,4'-dibromotriphenylamine;, 4,4'-dibromo-triphenylamine, bis 4-bromophenyl phenylamine, bis 4-bromophenyl phenylamine;
PubChem CID 13594707
IUPAC Name 4-bromo-N-(4-bromophenyl)-N-phenylaniline
SMILES C1=CC=C(C=C1)N(C2=CC=C(C=C2)Br)C3=CC=C(C=C3)Br

Specifications

Melting Point 85°C
Color White
Quantity 5 g
Formula Weight 403.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dibromotriphenylamine