accessibility menu, dialog, popup

UserName

CAS RN 8068-28-8

NaNaNaNaNaH3CCH3ONHNHSOOOOHNH3COHONHNHSOOOONHNHOHNOHNSOOONHOHNSOOONHOCH3CH3NHOH3CCH3HNONHSOOONHOH3CHO

CAS RN 8068-28-8

IUPAC Name: pentasodium [(3-{3-hydroxy-2-[2-(6-methyloctanamido)-4-[(sulfonatomethyl)amino]butanamido]butanamido}-3-{[3-(1-hydroxyethyl)-12,15-bis(2-methylpropyl)-2,5,8,11,14,17,20-heptaoxo-6,9,18-tris({2-[(sulfonatomethyl)amino]ethyl})-1,4,7,10,13,16,19-heptaazacyclotricosan-21-yl]carbamoyl}propyl)amino]methanesulfonate
Synonyms: pentasodium colistimethate
Molecular Weight (g/mol): 1749.81
Molecular Formula: C58H105N16Na5O28S5
InChi Key: IQWHCHZFYPIVRV-UHFFFAOYNA-I
SMILES: [Na+].[Na+].[Na+].[Na+].[Na+].CCC(C)CCCCC(=O)NC(CCNCS([O-])(=O)=O)C(=O)NC(C(C)O)C(=O)NC(CCNCS([O-])(=O)=O)C(=O)NC1CCNC(=O)C(NC(=O)C(CCNCS([O-])(=O)=O)NC(=O)C(CCNCS([O-])(=O)=O)NC(=O)C(CC(C)C)NC(=O)C(CC(C)C)NC(=O)C(CCNCS([O-])(=O)=O)NC1=O)C(C)O
Color 
  • (2)
Quantity 
  • (1)
  • (1)
Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)