missing translation for 'onlineSavingsMsg'
Learn More

Colistimethate Sodium Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015599
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015599 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015599 Supplier Merck Emd Millipore Supplier No. PHR1737500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Colistimethate Sodium belongs to the class of polymyxin antibiotics that acts by inhibiting the Gram negative bacteria. It is a prodrug that can hydrolyze to colistin in vivo or in vitro. It is employed as a last resort antibiotic to treat Pseudomonas aeruginosa infection in cystic fibrosis patients. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 8068-28-8
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Synonym Colistimethate sodium; Colistin sulphomethate sodium
Type Certified Reference Material
Quantity 500 mg
Grade Certified Reference Material