CAS RN 80676-35-3
IUPAC Name:
1-(chloromethyl)-4-(2-phenylethyl)benzene
Synonyms:
1-(chloromethyl)-4-(2-phenylethyl)benzene
Molecular Weight (g/mol):
230.74
Molecular Formula:
C15H15Cl
InChi Key:
KPTSLALCCGBCOB-UHFFFAOYSA-N
SMILES:
ClCC1=CC=C(CCC2=CC=CC=C2)C=C1
1
–
1
of
1
results
1
4-(Chloromethyl)-1,2-diphenylethane, 98%
CAS: 80676-35-3 Molecular Formula: C15H15Cl Molecular Weight (g/mol): 230.74 MDL Number: MFCD02093918 InChI Key: KPTSLALCCGBCOB-UHFFFAOYSA-N Synonym: 4-chloromethyl dibenzyl,1-chloromethyl-4-phenethylbenzene,4-chloromethyl-1,2-diphenylethane,1-chloromethyl-4-2-phenylethyl benzene,4-chloromethyl bibenzyl,4-phenethylbenzyl chloride PubChem CID: 2733990 IUPAC Name: 1-(chloromethyl)-4-(2-phenylethyl)benzene SMILES: ClCC1=CC=C(CCC2=CC=CC=C2)C=C1