missing translation for 'onlineSavingsMsg'
Learn More

4-(Chloromethyl)-1,2-diphenylethane, 98%

Catalog No. AAB2522214
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2522214 25 g
AAB2522203 1 g
AAB2522206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2522214 Supplier Thermo Scientific Chemicals Supplier No. B2522214
Only null left

CAS: 80676-35-3 | C15H15Cl | 230.74 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 80676-35-3
Molecular Formula C15H15Cl
Molecular Weight (g/mol) 230.74
MDL Number MFCD02093918
InChI Key KPTSLALCCGBCOB-UHFFFAOYSA-N
Synonym 4-chloromethyl dibenzyl, 1-chloromethyl-4-phenethylbenzene, 4-chloromethyl-1,2-diphenylethane, 1-chloromethyl-4-2-phenylethyl benzene, 4-chloromethyl bibenzyl, 4-phenethylbenzyl chloride
PubChem CID 2733990
IUPAC Name 1-(chloromethyl)-4-(2-phenylethyl)benzene
SMILES ClCC1=CC=C(CCC2=CC=CC=C2)C=C1

Specifications

Melting Point 38°C to 42°C
Quantity 25 g
UN Number UN1759
Formula Weight 230.73
Percent Purity 98%
Chemical Name or Material 4-(Chloromethyl)-1,2-diphenylethane
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only