accessibility menu, dialog, popup

UserName

CAS RN 80471-69-8

IUPAC Name: disodium 2-(2,7-dichloro-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
Synonyms: disodium 2-(2,7-dichloro-3-oxido-6-oxoxanthen-9-yl)benzoate
Molecular Weight (g/mol): 445.16
Molecular Formula: C20H8Cl2Na2O5
InChi Key: SZSVOWHDMGIUKH-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=C(Cl)C(=O)C=C2OC2=CC([O-])=C(Cl)C=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results