missing translation for 'onlineSavingsMsg'
Learn More

2',7'-Dichlorofluorescein Sodium Salt 98.0+%, TCI America™

Catalog No. D042425G
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D042425G 25 g
D04241G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D042425G Supplier TCI America Supplier No. D042425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 80471-69-8
Molecular Formula C20H8Cl2Na2O5
Molecular Weight (g/mol) 445.16
MDL Number MFCD00070511
InChI Key SZSVOWHDMGIUKH-UHFFFAOYSA-L
PubChem CID 10072180
IUPAC Name disodium 2-(2,7-dichloro-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
SMILES [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=C(Cl)C(=O)C=C2OC2=CC([O-])=C(Cl)C=C12

Specifications

Quantity 25 g
Formula Weight 445.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2′,7′-Dichlorofluorescein Sodium Salt