CAS RN 80360-14-1
IUPAC Name:
1-(benzenesulfonyl)-3-iodo-1H-indole
Synonyms:
1-(benzenesulfonyl)-3-iodoindole
Molecular Weight (g/mol):
383.2
Molecular Formula:
C14H10INO2S
InChi Key:
GKYWOZYEMLEJFK-UHFFFAOYSA-N
SMILES:
IC1=CN(C2=CC=CC=C12)S(=O)(=O)C1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
3-Iodo-1-(phenylsulfonyl)indole, 95%
3-Iodo-1-(phenylsulfonyl)indole, 95%
CAS: 80360-14-1 Molecular Formula: C14H10INO2S Molecular Weight (g/mol): 383.20 MDL Number: MFCD09037474 InChI Key: GKYWOZYEMLEJFK-UHFFFAOYSA-N Synonym: 3-iodo-1-phenylsulfonyl-1h-indole,3-iodo-1-phenylsulfonyl indole,1-benzenesulfonyl-3-iodo-1h-indole,1-benzenesulfonyl-3-iodoindole,1-benzenesulfonyl-3-iodo-indole,n-benzenesulfonyl-3-iodoindole,1-phenylsulfonyl-3-iodo-1h-indole,1h-indole,3-iodo-1-phenylsulfonyl PubChem CID: 10927077 IUPAC Name: 1-(benzenesulfonyl)-3-iodoindole SMILES: IC1=CN(C2=CC=CC=C12)S(=O)(=O)C1=CC=CC=C1