accessibility menu, dialog, popup

UserName

CAS RN 80220-63-9

IUPAC Name: (3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl)phosphonic acid
Synonyms: 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecylphosphonic acid
Molecular Weight (g/mol): 528.1
Molecular Formula: C10H6F17O3P
InChi Key: CETXMCMQEXPPLV-UHFFFAOYSA-N
SMILES: OP(O)(=O)CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results