missing translation for 'onlineSavingsMsg'
Learn More

(1H,1H,2H,2H-Heptadecafluorodecyl)phosphonic Acid 96.0+%, TCI America™

Catalog No. H1459200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H1459200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H1459200MG Supplier TCI America Supplier No. H1459200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 80220-63-9
Molecular Formula C10H6F17O3P
Molecular Weight (g/mol) 528.102
MDL Number MFCD19105612
InChI Key CETXMCMQEXPPLV-UHFFFAOYSA-N
Synonym (1H,1H,2H,2H-Perfluorodecyl)phosphonic Acid
PubChem CID 22630493
IUPAC Name 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecylphosphonic acid
SMILES C(CP(=O)(O)O)C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Melting Point 182°C
Color White
Quantity 200 mg
Formula Weight 528.10
Percent Purity ≥96.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (1H,1H,2H,2H-Heptadecafluorodecyl)phosphonic Acid