accessibility menu, dialog, popup

UserName

CAS RN 8009-03-8

IUPAC Name: —
Synonyms: petroleum jelly
Molecular Weight (g/mol): —
Molecular Formula: —
InChi Key: —
SMILES: CC1=NC2=CC=C3C=CC=CC3=C2C1(C)C
Grade 
  • (2)
  • (8)
Physical Form 
  • (4)
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (8)
  • (2)
  • (2)
  • (8)
  • (4)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)