missing translation for 'onlineSavingsMsg'
Learn More

Petrolatum, Yellow, USP, Spectrum™ Chemical

Catalog No. 18605054
Change view
Click to view available options
Quantity:
500 g
2.5 kg
15.9 kg
175 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605054 175 kg Poly Bottle
18605051 500 g Poly Bottle
18605052 2.5 kg Poly Bottle
18605053 15.9 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18605054 Supplier Spectrum Chemical Mfg Cor Supplier No. PE130175KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

PE130, 8009-03-8

Spectrum™ Chemical Petrolatum, Yellow, USP is also known as petroleum jelly and is a mixture of hydrocarbons. It is typically used as a topical ointment because of its healing properties and cosmetic purposes. The USP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 8009-03-8
Molecular Formula C15H15N
Molecular Weight (g/mol) 209.29
InChI Key WJZSZXCWMATYFX-UHFFFAOYSA-N
IUPAC Name 1,1,2-trimethyl-1H-benzo[e]indole
SMILES CC1=NC2=CC=C3C=CC=CC3=C2C1(C)C

Specifications

CAS Max % 1
Ignition Residue 0.001
Melting Range 38-60°C
Packaging Poly Bottle
Quantity 175 kg
Specific Gravity 0.815-0.880
Grade USP
Physical Form Powder or Waxy Solid
Chemical Name or Material Petrolatum, Yellow