accessibility menu, dialog, popup

UserName

CAS RN 80040-79-5

IUPAC Name: (2R,3R,4R)-3,4-bis(benzyloxy)-2-[(benzyloxy)methyl]-3,4-dihydro-2H-pyran
Synonyms: (2R,3R,4R)-3,4-bis(benzyloxy)-2-[(benzyloxy)methyl]-3,4-dihydro-2H-pyran
Molecular Weight (g/mol): 416.52
Molecular Formula: C27H28O4
InChi Key: MXYLLYBWXIUMIT-ZONZVBGPSA-N
SMILES: C(OCC1=CC=CC=C1)[C@H]1OC=C[C@@H](OCC2=CC=CC=C2)[C@H]1OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results