missing translation for 'onlineSavingsMsg'
Learn More

3,4,6-Tri-O-benzyl-D-galactal, Thermo Scientific Chemicals

Catalog No. AAJ6620206
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6620206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6620206 Supplier Thermo Scientific Chemicals Supplier No. J6620206

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 80040-79-5 | Molecular Formula: C27H28O4 | Molecular Weight: 416.52 g/mol

An important building block for solution- and solid-phase synthesis of oligosaccharides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 80040-79-5
Molecular Formula C27H28O4
Molecular Weight (g/mol) 416.52
MDL Number MFCD09039276
InChI Key MXYLLYBWXIUMIT-ZONZVBGPSA-N
Synonym tri-o-benzyl-d-galactal, 3,4,6-tri-o-benzyl-d-galactal, 2r,3r,4r-3,4-bis benzyloxy-2-benzyloxy methyl-3,4-dihydro-2h-pyran, 3-o,4-o,6-o-tribenzyl-1,2-dideoxy-d-lyxo-1-hexenopyranose, 1,5-anhydro-2-deoxy-d-lyxo-hex-1-enitol 3,4,6-tribenzyl ether, 2r,3r,4r-3,4-bis phenylmethoxy-2-phenylmethoxymethyl-3,4-dihydro-2h-pyran
PubChem CID 2734741
IUPAC Name (2R,3R,4R)-3,4-bis(benzyloxy)-2-[(benzyloxy)methyl]-3,4-dihydro-2H-pyran
SMILES C(OCC1=CC=CC=C1)[C@H]1OC=C[C@@H](OCC2=CC=CC=C2)[C@H]1OCC1=CC=CC=C1

Specifications

Melting Point 48°C to 54°C
Color White to Yellow
Flash Point >110°C (235°F)
Quantity 5 g
Solubility Information Soluble in dichloromethane or chloroform,
Optical Rotation −43° (c=1 in Dichloromethane)
Formula Weight 416.51
Physical Form Powder
Chemical Name or Material 3,4,6-Tri-O-benzyl-D-galactal
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only