CAS RN 80-71-7
CAS RN 80-71-7
IUPAC Name:
2-hydroxy-3-methylcyclopent-2-en-1-one
Synonyms:
methylcyclopentenolone
Molecular Weight (g/mol):
112.13
Molecular Formula:
C6H8O2
InChi Key:
CFAKWWQIUFSQFU-UHFFFAOYSA-N
SMILES:
CC1=C(O)C(=O)CC1
1
–
2
of
2
results
1
Methyl cyclopentenolone, ≥98% (GC), MilliporeSigma™ Supelco™
Methyl cyclopentenolone is an organic compound, commonly used in commercial products such as in E-liquids for electronic cigarettes.
2
2-Hydroxy-3-methyl-2-cyclopentenone 98.0+%, TCI America™
CAS: 80-71-7 Molecular Formula: C6H8O2 Molecular Weight (g/mol): 112.128 MDL Number: MFCD00013747 InChI Key: CFAKWWQIUFSQFU-UHFFFAOYSA-N Synonym: Cyclotene, 3-Methyl-1,2-cyclopentanedione, 3-Methyl-2-cyclopentenon-2-ol PubChem CID: 6660 IUPAC Name: 2-hydroxy-3-methylcyclopent-2-en-1-one SMILES: CC1=C(C(=O)CC1)O