missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxy-3-methyl-2-cyclopentenone 98.0+%, TCI America™
Supplier: TCI America H046925G
Specifications
| 2-Hydroxy-3-methyl-2-cyclopentenone | |
| 107°C | |
| MFCD00013747 | |
| Cyclotene, 3-Methyl-1,2-cyclopentanedione, 3-Methyl-2-cyclopentenon-2-ol | |
| CC1=C(C(=O)CC1)O | |
| 112.128 | |
| 112.13 | |
| Crystalline Powder |
| 80-71-7 | |
| C6H8O2 | |
| 25 g | |
| CFAKWWQIUFSQFU-UHFFFAOYSA-N | |
| 2-hydroxy-3-methylcyclopent-2-en-1-one | |
| 6660 | |
| ≥98.0% (T) |
Chemical Identifiers
| 80-71-7 | |
| 112.128 | |
| CFAKWWQIUFSQFU-UHFFFAOYSA-N | |
| 6660 | |
| CC1=C(C(=O)CC1)O |
| C6H8O2 | |
| MFCD00013747 | |
| Cyclotene, 3-Methyl-1,2-cyclopentanedione, 3-Methyl-2-cyclopentenon-2-ol | |
| 2-hydroxy-3-methylcyclopent-2-en-1-one |
Safety and Handling
EINECSNumber : (3)-2406
RTECSNumber : GY7298000
TSCA : Yes