missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl cyclopentenolone, ≥98% (GC), MilliporeSigma™ Supelco™
Methyl cyclopentenolone is an organic compound, commonly used in commercial products such as in E-liquids for electronic cigarettes.
Supplier: Merck Emd Millipore 80133100MG
Specifications
| 80-71-7 | |
| C6H8O2 | |
| Cyclotene | |
| 100 mg | |
| ≥98% (GC) |
| Neat | |
| 104°C to 108°C; 104°C to 108°C | |
| Analytical Standard | |
| 112.13 | |
| Analytical Standard |