accessibility menu, dialog, popup

UserName

CAS RN 80-43-3

IUPAC Name: {2-[(2-phenylpropan-2-yl)peroxy]propan-2-yl}benzene
Synonyms: {2-[(2-phenylpropan-2-yl)peroxy]propan-2-yl}benzene
Molecular Weight (g/mol): 270.37
Molecular Formula: C18H22O2
InChi Key: XMNIXWIUMCBBBL-UHFFFAOYSA-N
SMILES: CC(C)(OOC(C)(C)C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)