missing translation for 'onlineSavingsMsg'
Learn More

Dicumyl Peroxide (contains ca. 60% CaCO3), TCI America™

Catalog No. D4894100G
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4894100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4894100G Supplier TCI America Supplier No. D4894100G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 80-43-3
Molecular Formula C18H22O2
Molecular Weight (g/mol) 270.372
MDL Number MFCD00036227
InChI Key XMNIXWIUMCBBBL-UHFFFAOYSA-N
Synonym dicumyl peroxide, cumene peroxide, cumyl peroxide, percumyl d, perkadox b, diisopropylbenzene peroxide, perkadox bc, perkadox sb, dicumenyl peroxide, peroxide, bis 1-methyl-1-phenylethyl
PubChem CID 6641
IUPAC Name 2-(2-phenylpropan-2-ylperoxy)propan-2-ylbenzene
SMILES CC(C)(C1=CC=CC=C1)OOC(C)(C)C2=CC=CC=C2

Specifications

Color White
Quantity 100 g
UN Number 3077
Formula Weight 270.37
Physical Form Crystalline Powder
Chemical Name or Material Dicumyl Peroxide (contains ca. 60% CaCO3)
EINECSNumber (3)-1086
RTECSNumber SD8150000
TSCA Yes
Recommended Storage Refrigerator