accessibility menu, dialog, popup

UserName

NBrBrFFF

CAS RN 79623-38-4

IUPAC Name: 2,3-dibromo-5-(trifluoromethyl)pyridine
Synonyms: 2,3-dibromo-5-(trifluoromethyl)pyridine
Molecular Weight (g/mol): 304.89
Molecular Formula: C6H2Br2F3N
InChi Key: BIDFUURRDIAINQ-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(Br)=C(Br)N=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results