missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dibromo-5-(trifluoromethyl)pyridine, 95%, Thermo Scientific Chemicals

Catalog No. AAH61407MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH61407MD 250 mg
AAH6140703 1 g
AAH6140706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH61407MD Supplier Thermo Scientific Chemicals Supplier No. H61407MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 79623-38-4
Molecular Formula C6H2Br2F3N
Molecular Weight (g/mol) 304.89
MDL Number MFCD10687233
InChI Key BIDFUURRDIAINQ-UHFFFAOYSA-N
PubChem CID 26985667
IUPAC Name 2,3-dibromo-5-(trifluoromethyl)pyridine
SMILES FC(F)(F)C1=CC(Br)=C(Br)N=C1

Specifications

Quantity 250 mg
Synonym 2,3-dibromo-5-trifluoromethyl pyridine, pyridine, 2,3-dibromo-5-trifluoromethyl, pyridine,2,3-dibromo-5-trifluoromethyl, pubchem15128, acmc-209phs, 2, 3-dibromo-5-trifluoromethyl pyridine
Formula Weight 304.89
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2,3-Dibromo-5-(trifluoromethyl)pyridine
Hazard Category H302+H312+H332-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only