accessibility menu, dialog, popup

UserName

CAS RN 79404-93-6

IUPAC Name: 2,4,5-trichlorophenyl 4-(octyloxy)benzoate
Synonyms: 2,4,5-trichlorophenyl 4-(octyloxy)benzoate
Molecular Weight (g/mol): 429.76
Molecular Formula: C21H23Cl3O3
InChi Key: ILQVFGUXJOSAJH-UHFFFAOYSA-N
SMILES: CCCCCCCCOC1=CC=C(C=C1)C(=O)OC1=CC(Cl)=C(Cl)C=C1Cl
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results