missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trichlorophenyl 4-n-Octyloxybenzoate, TCI America™

Catalog No. O023025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O023025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O023025G Supplier TCI America Supplier No. O023025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 79404-93-6
Molecular Formula C21H23Cl3O3
Molecular Weight (g/mol) 429.762
MDL Number MFCD00191565
InChI Key ILQVFGUXJOSAJH-UHFFFAOYSA-N
Synonym 4-n-Octyloxybenzoic Acid 2,4,5-Trichlorophenyl Ester
PubChem CID 616208
IUPAC Name (2,4,5-trichlorophenyl) 4-octoxybenzoate
SMILES CCCCCCCCOC1=CC=C(C=C1)C(=O)OC2=CC(=C(C=C2Cl)Cl)Cl

Specifications

Melting Point 56°C
Color White
Quantity 25 g
Formula Weight 429.76
Physical Form Crystalline Powder
Chemical Name or Material 2,4,5-Trichlorophenyl 4-n-Octyloxybenzoate