accessibility menu, dialog, popup

UserName

HOSi

CAS RN 791-31-1

IUPAC Name: triphenylsilanol
Synonyms: triphenylsilanol 1,1,1-triphenyl-silanol silane, (hydroxytriphenyl)- C18H16osi Si(OH)Ph3 (hydroxytriphenyl)-silane silanol, 1,1,1-triphenyl- silanol, triphenyl-
Molecular Weight (g/mol): 276.41
Molecular Formula: C18H16OSi
InChi Key: NLSXASIDNWDYMI-UHFFFAOYSA-N
SMILES: O[Si](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)