accessibility menu, dialog, popup

UserName

CAS RN 787-84-8

OHNNHO

CAS RN 787-84-8

IUPAC Name: N'-benzoylbenzohydrazide
Synonyms: dibenzoylhydrazine C6H5-CO-NH-NH-CO-C6H5
Molecular Weight (g/mol): 240.26
Molecular Formula: C14H12N2O2
InChi Key: GRRIYLZJLGTQJX-UHFFFAOYSA-N
SMILES: O=C(NNC(=O)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results