missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N'-Dibenzoylhydrazine 95.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 240.26 |
| Percent Purity | ≥95.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | N,N′-Dibenzoylhydrazine |
Safety and Handling
| RTECSNumber | MV2085000 |
| TSCA | Yes |