accessibility menu, dialog, popup

UserName

HOOOHO

CAS RN 787-70-2

IUPAC Name: [1,1'-biphenyl]-4,4'-dicarboxylic acid
Synonyms: santalum album santalum album wood powder santalum album wood oil santalum album extract santalum album seed extract santalum album wood extract
Molecular Weight (g/mol): 242.23
Molecular Formula: C14H10O4
InChi Key: NEQFBGHQPUXOFH-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1)C1=CC=C(C=C1)C(O)=O
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)