missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Biphenyldicarboxylic Acid 97.0+%, TCI America™

Catalog No. B11915G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11915G 5 g
B119125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B11915G Supplier TCI America Supplier No. B11915G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 787-70-2
Molecular Formula C14H10O4
Molecular Weight (g/mol) 242.23
MDL Number MFCD00002554
InChI Key NEQFBGHQPUXOFH-UHFFFAOYSA-N
Synonym biphenyl-4,4'-dicarboxylic acid, 4,4'-biphenyldicarboxylic acid, 1,1'-biphenyl-4,4'-dicarboxylic acid, 4-4-carboxyphenyl benzoic acid, 4,4'-bibenzoic acid, 4,4'-diphenic acid, 4,4'-dicarboxybiphenyl, p,p'-diphenic acid, usaf do-69, bibenzoic acid
PubChem CID 13084
IUPAC Name 4-(4-carboxyphenyl)benzoic acid
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)C(=O)O)C(=O)O

Specifications

Quantity 5 g
Formula Weight 242.23
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Biphenyldicarboxylic Acid
RTECSNumber DV2975000
TSCA Yes