accessibility menu, dialog, popup

UserName

CAS RN 78618-06-1

IUPAC Name: benzyl N-(6-aminohexyl)carbamate hydrochloride
Synonyms: benzyl N-(6-aminohexyl)carbamate hydrochloride
Molecular Weight (g/mol): 286.8
Molecular Formula: C14H23ClN2O2
InChi Key: GMSZGOGHNFZOMF-UHFFFAOYSA-N
SMILES: Cl.NCCCCCCNC(=O)OCC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)