missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxy-1,6-diaminohexane Hydrochloride 98.0+%, TCI America™

Catalog No. C15215G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15215G 5 g
C15211G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C15215G Supplier TCI America Supplier No. C15215G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 78618-06-1
Molecular Formula C14H23ClN2O2
Molecular Weight (g/mol) 286.8
MDL Number MFCD00270151
InChI Key GMSZGOGHNFZOMF-UHFFFAOYSA-N
Synonym N-(6-Aminohexyl)carbamic Acid Benzyl Ester Hydrochloride, Benzyl N-(6-Aminohexyl)carbamate Hydrochloride, N-Cbz-1,6-diaminohexane Hydrochloride
PubChem CID 13196229
IUPAC Name benzyl N-(6-aminohexyl)carbamate;hydrochloride
SMILES C1=CC=C(C=C1)COC(=O)NCCCCCCN.Cl

Specifications

Melting Point 177°C
Color White-Yellow
Quantity 5 g
Formula Weight 286.80
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-1,6-diaminohexane Hydrochloride