accessibility menu, dialog, popup

UserName

CAS RN 78607-32-6

IUPAC Name: 2,5-dichloropyridin-3-amine
Synonyms: 2,5-dichloropyridin-3-amine
Molecular Weight (g/mol): 163
Molecular Formula: C5H4Cl2N2
InChi Key: OLZPJUVEGSNIJL-UHFFFAOYSA-N
SMILES: NC1=CC(Cl)=CN=C1Cl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results